Order Entry
Italy
ContactUsLinkComponent
 
2-(3-Thienyl)-1,3-dioxolane ≥97.0% (by GC)
  TCIAT2623-5G
 :  TCI
 :  
2-(3-Thienyl)-1,3-dioxolane ≥97.0% (by GC)
  TCIAT2623-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :156.20 g/mol
 :84°C
 :Refrigerator
 :13250-82-3
 :C₇H₈O₂S
 :MFCD06209359