
Thieno[3,2-b]thiophene-2-boronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2621-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
About this item
:
:184.03 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:160032-40-6
Formula:C₆H₅BO₂S₂
MDL Number:MFCD08059468