



Trimethylsilylmethylmagnesium chloride ∼18% ∼1 mol/L in THF
Catalog # TCIAT2609-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Concentration (Neutralization titration):18.5 to 20.0 w/w%
Specification Test Results
About this item
:
:146.97 g/mol
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:13170-43-9
Formula:C₄H₁₁ClMgSi
MDL Number:MFCD00009916