Order Entry
Italy
ContactUsLinkComponent
p-Tolyl trifluoromethanesulphonate ≥95.0% (by GC)
p-Tolyl trifluoromethanesulphonate ≥95.0% (by GC)
  TCIAT2606-5G
 :  TCI
 :  
p-Tolyl trifluoromethanesulphonate ≥95.0% (by GC)
  TCIAT2606-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :240.20 g/mol
 :115°C
 :89°C
 :GHS07
 :29540-83-8
 :C₈H₇F₃O₃S
 :MFCD00192507