
O-(Tetrahydropyran-2-yl)-hydroxylamine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6723-30-4
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD01321374
:117.148 g/mol
Boiling Pt:81°C
Melting Pt:36°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together