
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:128055-74-3
:
Formula:C₂₅H₁₂Br₄
MDL Number:MFCD08704220
:631.9800 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together