Order Entry
Italy
ContactUsLinkComponent
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥97.0% (by GC)
Irritant
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥97.0% (by GC)
Catalog # TCIAT2540-200MG
Supplier:  TCI
CAS Number:  
Irritant
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥97.0% (by GC)
Catalog # TCIAT2540-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    85.0 to 89.0 deg-C

Specification Test Results

About this item

 :
 :544.69 g/mol
 :GHS07
CAS Number:126320-57-8
Formula:C₂₆H₄₈N₄O₈
MDL Number:MFCD09263316