
Tizanidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:64461-82-1
:
Formula:C₉H₈ClN₅S
MDL Number:MFCD00798231
:290.17 g/mol
Merck Index:14,09485
Melting Pt:280°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together