Order Entry
Italy
ContactUsLinkComponent
Thiourea ≥99.0% (by titrimetric analysis)
Irritant
Health
Environment
Thiourea ≥99.0% (by titrimetric analysis)
Catalog # TCIAT2475-300G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
Environment
Thiourea ≥99.0% (by titrimetric analysis)
Catalog # TCIAT2475-300G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    300 g
  • Seq:
    3575

Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Iodometric back titration):
    min. 99.0 %

Specification Test Results

About this item

 :
 :76.12 g/mol
Merck Index:14,09367
Melting Pt:177°C
EINECS:200-543-5
UN:2811
ADR:6.1,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:62-56-6
Formula:NH₂CSNH₂
MDL Number:MFCD00008067