
N-(3-Trifluoromethylphenyl)anthranilic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT2354-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
Specification Test Results
About this item
:
:281.23 g/mol
Merck Index:14,04132
EINECS:208-494-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:530-78-9
Formula:C₁₄H₁₀F₃NO₂
MDL Number:MFCD00002422