D-ɑ-Tocopherylquinone ≥97.0% (by HPLC)
Catalog # TCIAT2283-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:446.72 g/mol
Storage Temperature:Refrigerator
EINECS:231-450-8
CAS Number:7559-04-8
Formula:C₂₉H₅₀O₃
MDL Number:MFCD00135657