Order Entry
Italy
ContactUsLinkComponent
 
Tetradecyl lactate ≥75.0% (by GC)
  TCIAT2231-25G
 :  TCI
 :  
Tetradecyl lactate ≥75.0% (by GC)
  TCIAT2231-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 75.0 %

 

 

 : Tetradecyl 2-Hydroxypropionate, Lactic Acid Tetradecyl Ester, Myristyl Lactate
 :
 :286.46 g/mol
 :215-350-1
 :1323-03-1
 :C₁₇H₃₄O₃