1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-β-D-glucopyranose ≥97.0% (by HPLC)
: TCI
:80321-89-7
:
:C₁₄H₁₉N₃O₉
:373.32 g/mol
:Freezer
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:95.0 to 99.0 deg-C
- Specific rotation [a]20/D:+2.5 to +3.8 deg (C=1, CHCl3)
Frequently Bought Together