
O,O'-Di-2-pyridyl thiocarbonate ≥98.0% (by total nitrogen basis)
Catalog # TCIAT1906-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:97.0 to 102.0 deg-C
Specification Test Results
About this item
:
:232.26 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:96989-50-3
Formula:C₁₁H₈N₂O₂S
MDL Number:MFCD00074870