Order Entry
Italy
ContactUsLinkComponent
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥97.0% (by titrimetric analysis)
Corrosive
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Corrosive
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    87.0 to 90.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

Frequently Bought Together