Order Entry
Italy
ContactUsLinkComponent
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥98.0% (by titrimetric analysis)
 
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥98.0% (by titrimetric analysis)
  TCIAT1901-1G
 :  TCI
 :  
 
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥98.0% (by titrimetric analysis)
  TCIAT1901-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    108.0 to 112.0 deg-C
  • Specific rotation [a]20/D:
    -16.0 to -17.0 deg (C=5, H2O)
  • Solubility in Water:
    almost transparency

 

 

 :
 :158.08 g/mol
 :Refrigerator
 :3261
 :8,III
 :GHS05
 :24435-45-8
 :C₄H₅F₃O₃
 :MFCD03092898