Order Entry
Italy
ContactUsLinkComponent
 
 
6-Bromo-2-naphthyl trifluoromethanesulphonate ≥95.0% (by GC)
 :  TCI
 
 
6-Bromo-2-naphthyl trifluoromethanesulphonate ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    52.0 to 56.0 deg-C
  • Solubility in Toluene:
    very faint turbidity

 

Frequently Bought Together