Order Entry
Italy
ContactUsLinkComponent
Trinonylamine ≥98.0% (by titrimetric analysis)
Irritant
Trinonylamine ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1826-5ML
Supplier:  TCI
CAS Number:  
Irritant
Trinonylamine ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1826-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :395.76 g/mol
Boiling Pt:270°C
 :GHS07
CAS Number:2044-22-6
Formula:C₂₇H₅₇N
MDL Number:MFCD00053817