
Tetracyclo[6.2.1.1(3,6).0(2,7)]dodec-4-ene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:21635-90-5
:
Formula:C₁₂H₁₆
MDL Number:MFCD00213460
:160.26 g/mol
Boiling Pt:93°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together