6,6',7,7'-Tetrahydroxy-4,4,4',4'-tetramethyl-2,2'-spirobichroman ≥98.0% (by HPLC)
Catalog # TCIAT1791-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:372.42 g/mol
CAS Number:32737-35-2
Formula:C₂₁H₂₄O₆
MDL Number:MFCD00627043