
2,2',3,3'-Tetrahydroxy-1,1'-binaphthyl ≥98.0% (by HPLC)
TCIAT1754-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
:
:318.33 g/mol
:GHS07
:39215-21-9
:C₂₀H₁₄O₄
:MFCD00188003