Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III) ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17904-83-5
:
Formula:C₅₇H₄₁EuN₂O₆
MDL Number:MFCD01321202
:1001.93 g/mol
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
- Melting point:186.0 to 192.0 deg-C
Specification Test Results
Frequently Bought Together