Dimethyl tetradecanedioate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5024-21-5
:
Formula:C₁₆H₃₀O₄
MDL Number:MFCD00673459
:286.41 g/mol
Boiling Pt:196°C
EINECS:225-711-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:43.0 to 46.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together