O-(2-Trimethylsilylethyl)hydroxylamine Hydrochloride ≥98.0% (by total nitrogen basis)
Catalog # TCIAT1725-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:169.72 g/mol
CAS Number:153502-27-3
Formula:C₅H₁₅NOSi
MDL Number:MFCD01321199