Order Entry
Italy
ContactUsLinkComponent
Hafnium(IV) trifluoromethanesulphonate ≥90.0% (by titrimetric analysis)
Corrosive
Hafnium(IV) trifluoromethanesulphonate ≥90.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Corrosive
Hafnium(IV) trifluoromethanesulphonate ≥90.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Chelometric Titration):
    min. 90.0 %

Specification Test Results

Related Information

Frequently Bought Together