



p-Tolylmagnesium bromide 19% ∼1 mol/L in THF
Catalog # TCIAT1700-100G
Supplier: TCI
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Concentration (sec Butanol method):19.0 to 20.9 %
Specification Test Results
About this item
:
:195.34 g/mol
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:4294-57-9
Formula:C₇H₇BrMg
MDL Number:MFCD00000040