
Tri(2-thienyl)phosphine ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24171-89-9
:
Formula:C₁₂H₉PS₃
MDL Number:MFCD00015471
:280.36 g/mol
Boiling Pt:205°C
EINECS:246-059-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Amber to Dark green powder to lump
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together