
Titanocene bis(trifluoromethanesulphonate) ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:76262-87-8
:
Formula:C₁₂H₁₀F₆O₆S₂Ti
MDL Number:MFCD00192505
:476.18 g/mol
:GHS07
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (Chelometric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together