Order Entry
Italy
ContactUsLinkComponent
2,2'-Thiobis(ethylamine) ≥98.0% (by titrimetric analysis)
Corrosive
2,2'-Thiobis(ethylamine) ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1538-10G
Supplier:  TCI
CAS Number:  
Corrosive
2,2'-Thiobis(ethylamine) ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1538-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :120.21 g/mol
Boiling Pt:120°C
EINECS:212-810-3
UN:2735
ADR:8,III
 :GHS05
CAS Number:871-76-1
Formula:C₄H₁₂N₂S
MDL Number:MFCD00039783