Order Entry
Italy
ContactUsLinkComponent
2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (by GC)
Irritant
2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (by GC)
Catalog # TCIAT1522-1G
Supplier:  TCI
CAS Number:  
Irritant
2,2,2-Trifluoro-1-(9-anthryl)ethanol ≥99.0% (by GC)
Catalog # TCIAT1522-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    142.0 to 144.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :276.26 g/mol
EINECS:265-791-9
 :GHS07
CAS Number:65487-67-4
Formula:C₁₆H₁₁F₃O
MDL Number:MFCD00062967