
1,4,5,6-Tetrahydro-5,6-dioxo-2,3-pyrazinedicarbonitrile ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1472-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
:162.11 g/mol
:GHS07
CAS Number:36023-64-0
Formula:C₆H₂N₄O₂
MDL Number:MFCD00191690