Order Entry
Italy
ContactUsLinkComponent
 
N-sulphinyl-p-toluenesulphonamide ≥70% (by titrimetric analysis)
 :  TCI
 
undefined
N-sulphinyl-p-toluenesulphonamide ≥70% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (Iodometric Titration):
    min. 70.0 %

 

Frequently Bought Together