Order Entry
Italy
ContactUsLinkComponent
Trimethylsilylmethylmagnesium chloride 20% ∼1 mol/L in ethyl ether
Flammable
Corrosive
Irritant
Health
Trimethylsilylmethylmagnesium chloride 20% ∼1 mol/L in ethyl ether
Catalog # TCIAT1451-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Trimethylsilylmethylmagnesium chloride 20% ∼1 mol/L in ethyl ether
Catalog # TCIAT1451-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Concentration (Neutralization titration):
    20.0 to 22.0 w/w%

Specification Test Results

About this item

 :
 :146.97 g/mol
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:13170-43-9
Formula:C₄H₁₁ClMgSi
MDL Number:MFCD00009916