Trigonelline hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1429-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
Merck Index:14,09694
:173.60 g/mol
EINECS:228-119-5
CAS Number:6138-41-6
Formula:C₇H₈ClNO₂
MDL Number:MFCD00077250