Order Entry
Italy
ContactUsLinkComponent
5,6,7,8-Tetrahydro-2-methylquinoline ≥98.0% (by GC, titration analysis)
5,6,7,8-Tetrahydro-2-methylquinoline ≥98.0% (by GC, titration analysis)
  TCIAT1408-10ML
 :  TCI
 :  
undefined
5,6,7,8-Tetrahydro-2-methylquinoline ≥98.0% (by GC, titration analysis)
  TCIAT1408-10ML
 :  TCI
 :  

 

  • Pk:
    10 mL
  • Seq:
    2275

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :147.22 g/mol
 :225°C
 :GHS07
 :2617-98-3
 :C₁₀H₁₃N
 :MFCD00191581