



Tetrahexylammonium hydroxide 10% in methanol
Catalog # TCIAT1134-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
Specification Test Results
About this item
:
:371.69 g/mol
Flash Pt:11°C
UN:2924
ADR:3,III
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:17756-56-8
Formula:C₂₄H₅₃NO
MDL Number:MFCD00059255