Order Entry
Italy
ContactUsLinkComponent
Tetrakis(hydroxymethyl)phosphonium sulphate ∼70 - 80% in water
Corrosive
Toxic
Tetrakis(hydroxymethyl)phosphonium sulphate ∼70 - 80% in water
Supplier:  TCI
Certificates
Corrosive
Toxic
Tetrakis(hydroxymethyl)phosphonium sulphate ∼70 - 80% in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Iodometric titration):
    70.0 to 80.0 w/w%

Specification Test Results

Frequently Bought Together