



3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% in methanol for transesterification of glyceride
Supplier: TCI
Certificates
About this item
CAS Number:68254-41-1
:
Formula:C₁₀H₁₄F₃NO
MDL Number:MFCD00059473
:221.22 g/mol
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):5.0 to 5.5 w/w%
- High boiling impurities:to pass test
Specification Test Results
Related Information
Frequently Bought Together