

N,N,N',N'-Tetramethyl-1,2-diaminopropane ≥92.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1822-45-3
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00014871
:130.24 g/mol
Boiling Pt:72°C
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 92.0 %
Specification Test Results
Frequently Bought Together