Bis(2-ethylhexyl)-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2915-49-3
:
Formula:C₂₄H₄₂O₄
MDL Number:MFCD00059565
:394.60 g/mol
Flash Pt:202°C
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min.97.0 % (total of isomers)
Specification Test Results
Frequently Bought Together