
1,2,3-Tris(2-cyanoethoxy)propane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2465-93-2
:
Formula:C₁₂H₁₇N₃O₃
MDL Number:MFCD00019868
:251.29 g/mol
EINECS:219-573-5
UN:3276
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together