Order Entry
Italy
ContactUsLinkComponent
3,3',5-Triiodo-L-thyronine ≥98.0% (by HPLC, titration analysis)
Irritant
3,3',5-Triiodo-L-thyronine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT0453-100MG
Supplier:  TCI
CAS Number:  
Irritant
3,3',5-Triiodo-L-thyronine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT0453-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +21.0 to +25.0 deg (C=5, 1mol/L HCl:EtOH=1:2)

Specification Test Results

About this item

 :
 :650.98 g/mol
Merck Index:14,05510
EINECS:229-999-3
 :GHS07
CAS Number:6893-02-3
Formula:C₁₅H₁₂I₃NO₄
MDL Number:MFCD00002593