Order Entry
Italy
ContactUsLinkComponent
 
 
 
o-Tolidine solution 0.1% (w/v) in hydrochloric acid 2M
  TCIAT0255-500ML
 :  TCI
 :  
undefined
 
 
o-Tolidine solution 0.1% (w/v) in hydrochloric acid 2M
  TCIAT0255-500ML
 :  TCI
 :  

 

  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance:
    Colorless clear liquid
  • Sensitiveness:
    to pass test

 

 

 :
 :212.29 g/mol
 :3265
 :8,III
 :GHS05
 :GHS08
 :119-93-7
 :[-C₆H₃(CH₃)-₄-NH₂]₂
 :MFCD00014773