Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Tetradecyl ether ≥97.0% (by GC)

Tetradecyl ether ≥97.0% (by GC)

25 g
Catalog # TCIAT0091-25G
Supplier:   TCI
Tetradecyl ether ≥97.0% (by GC)
Catalog # TCIAT0091-25G
Supplier:  TCI
CAS Number:  5412-98-6
Tetradecyl ether ≥97.0% (by GC)

25 g

CAS Number:  5412-98-6
Price
144,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Ditetradecyl Ether, Dimyristyl Ether, Myristyl Ether
  • CAS Number:5412-98-6
  • Formula:C₂₈H₅₈O
  • MDL Number:MFCD00026607
  • Molecular Weight:410.77 g/mol
  • EINECS:226-499-7
Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    43.0 to 46.0 deg-C

Specifications

  • Conf:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.