Order Entry
Italy
ContactUsLinkComponent
2-(9,9'-Spirobi[fluoren]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9'-Spirobi[fluoren]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
2-(9,9'-Spirobi[fluoren]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    216.0 to 220.0 deg-C

Specification Test Results

Frequently Bought Together