Order Entry
Italy
ContactUsLinkComponent
Silver(I) trifluoromethanethiolate ≥95.0% (by titrimetric analysis)
Silver(I) trifluoromethanethiolate ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Silver(I) trifluoromethanethiolate ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (Precipitation Titration):
    min. 95.0 %

Specification Test Results

Frequently Bought Together