Order Entry
Italy
ContactUsLinkComponent
(R)-2,2',3,3'-Tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥98.0% (by GC)
(R)-2,2',3,3'-Tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥98.0% (by GC)
Catalog # TCIAS0957-1G
Supplier:  TCI
CAS Number:  
(R)-2,2',3,3'-Tetrahydro-1,1'-spirobi[indene]-7,7'-diol ≥98.0% (by GC)
Catalog # TCIAS0957-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %
  • Optical purity (LC):
    min. 98.0 ee%
  • Melting point:
    156.0 to 160.0 deg-C

Specification Test Results

About this item

 :
 :252.31 g/mol
CAS Number:223259-62-9
 :MFCD16621090
Formula:C₁₇H₁₆O₂