Order Entry
Italy
ContactUsLinkComponent
 
 
Sodium bromodifluoroacetate ≥98.0% (by HPLC, titration analysis)
  TCIAS0908-5G
 :  TCI
 :  
undefined
 
Sodium bromodifluoroacetate ≥98.0% (by HPLC, titration analysis)
  TCIAS0908-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

 

 

 :
 :196.91 g/mol
 :GHS07
 :84349-27-9
 :C₂HBrF₂O₂
 :MFCD04038355