Order Entry
Italy
ContactUsLinkComponent
4-Sulpho-1,8-naphthalic anhydride potassium salt ≥98.0% (by HPLC)
Irritant
4-Sulpho-1,8-naphthalic anhydride potassium salt ≥98.0% (by HPLC)
Catalog # TCIAS0904-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Sulpho-1,8-naphthalic anhydride potassium salt ≥98.0% (by HPLC)
Catalog # TCIAS0904-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Methanolysis Method):
    min. 95.0 %

Specification Test Results

About this item

 :
 :316.32 g/mol
EINECS:275-547-3
 :GHS07
CAS Number:71501-16-1
Formula:C₁₂H₆O₆S
MDL Number:MFCD00012097