
N-Succinimidyl-11-maleimidoundecanoate ≥98.0% (by HPLC)
Catalog # TCIAS0882-20MG
Supplier: TCI
CAS Number:
Specifications
- Pk:20 mg
- Seq:2305
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):7.1 to 7.5 %
- Melting point:90.0 to 94.0 deg-C
Specification Test Results
About this item
:
:378.43 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:87981-04-2
Formula:C₁₉H₂₆N₂O₆
MDL Number:MFCD00467873